Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isopropylpiperazine 97%

Supplier: AMBEED, INC
Danger

A133041-10G A133041-1G A133041-250MG A133041-25G A133041-5G
77109-360EA 17.52 USD InStock
77109-360 77109-362 77109-364 77109-366 77109-368
1-Isopropylpiperazine 97%
1-Isopropylpiperazine
Precaución: for Research Use Only


Formula: C₇H₁₆N₂
MDL Number: MFCD00167971
CAS Number: 4318-42-7
UN: 1992
ADR: 3,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty