Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl benzylcarbamate 95%

Supplier: AMBEED, INC
Warning

A134368-100MG A134368-1G A134368-250MG
77109-434EA 45.33 USD InStock
77109-434 77109-436 77109-438
Methyl benzylcarbamate 95%
Methyl benzylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MDL Number: MFCD00025856
CAS Number: 5817-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty