Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxypyridin-3-ol 95%

Supplier: AMBEED, INC
Warning

A135454-1G A135454-250MG A135454-25G A135454-5G
77109-492EA 19.57 USD InStock
77109-492 77109-494 77109-496 77109-498
2-Methoxypyridin-3-ol 95%
2-Methoxypyridin-3-ol
Precaución: for Research Use Only


Formula: C₆H₇NO₂
MDL Number: MFCD00457990
CAS Number: 13472-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty