Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: AMBEED, INC
Warning

A137797-10G A137797-1G A137797-25G A137797-5G
77109-648EA 192.61 USD InStock
77109-648 77109-650 77109-652 77109-654
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11046240
CAS Number: 876343-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty