Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-2-(1-methylethyl)-piperazine 97%

Supplier: AMBEED, INC
Warning

A137986-100MG A137986-1G A137986-1MG A137986-250MG A137986-5G
77109-666EA 16.47 USD InStock
77109-666 77109-668 77109-670 77109-672 77109-674
1-Boc-2-(1-methylethyl)-piperazine 97%
1-Boc-2-(1-methylethyl)-piperazine
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD08273957
CAS Number: 886766-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty