Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenylhydrazine hydrochloride 98%

Supplier: AMBEED, INC
Danger

A140593-10G A140593-25G A140593-5G
77109-788EA 51.99 USD InStock
77109-788 77109-790 77109-792 77109-794
4-Nitrophenylhydrazine hydrochloride 98%
4-Nitrophenylhydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
MDL Number: MFCD00040592
CAS Number: 636-99-7
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty