Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Glu-OAll 97%

Supplier: AMBEED, INC
Warning

A144458-1G A144458-250MG A144458-25G
77109-950EA 20.6 USD InStock
77109-950 77109-952 77109-954
Fmoc-D-Glu-OAll 97%
Fmoc-D-Glu-OAll
Precaución: for Research Use Only


Formula: C₂₃H₂₃NO₆
MW: 409.4 g/mol
MDL Number: MFCD01074691
CAS Number: 204251-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty