Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trifluorobenzoic acid 96%

Supplier: AMBEED, INC
Warning

A146972-100G A146972-10G A146972-25G A146972-5G
77110-070EA 51.5 USD InStock
77110-070 77110-072 77110-074 77110-076
2,4,5-Trifluorobenzoic acid 96%
2,4,5-Trifluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃F₃O₂
MDL Number: MFCD00013306
CAS Number: 446-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty