Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-1,3-cyclohexanedione 97%

Supplier: AMBEED, INC
Warning

A148361-10G A148361-1G A148361-5G A148361-250MG
77110-144EA 160.68 USD InStock
77110-144 77110-146 77110-148 77685-245
2-Acetyl-1,3-cyclohexanedione 97%
2-Acetyl-1,3-cyclohexanedione
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MDL Number: MFCD00001586
CAS Number: 4056-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty