Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SD208 98%

Supplier: AMBEED, INC
Warning

A150813-100MG A150813-10MG A150813-1MG A150813-50MG
77110-286EA 119.47 USD InStock
77110-286 77110-288 77110-290 77685-255
SD208 98%
SD208
Precaución: for Research Use Only


Formula: C₁₇H₁₀ClFN₆
MDL Number: MFCD11519969
CAS Number: 627536-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty