Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Aminobutanoic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A154015-100G A154015-1G A154015-5G A154015-25G
77110-428EA 89.61 USD InStock
77110-428 77110-430 77110-432 77685-276
(S)-2-Aminobutanoic acid hydrochloride 95%
(S)-2-Aminobutanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD02092984
CAS Number: 5959-29-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty