Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloropicolinic acid 98%

Supplier: AMBEED, INC
Warning

A156432-100G A156432-10G A156432-1G A156432-25G
77110-498EA 159.64 USD InStock
77110-498 77110-500 77110-502 77110-504
5-Chloropicolinic acid 98%
5-Chloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₄ClNO₂
MW: 157.56
MDL Number: MFCD04114192
CAS Number: 86873-60-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty