Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Aminoacetophenone 97%

Supplier: AMBEED, INC
Warning

A156836-100G A156836-10G A156836-500G A156836-5G A156836-25G A156836-1000G
77110-520EA 22.66 USD InStock
77110-520 77110-522 77110-524 77110-526 77318-936 77685-293
2'-Aminoacetophenone 97%
2'-Aminoacetophenone
Precaución: for Research Use Only


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 252
Melting Pt: 20
MDL Number: MFCD00007717
CAS Number: 551-93-9
Merck Index: 13,00412
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty