Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Iodoacetanilide 95%

Supplier: AMBEED, INC
Warning

A164324-100G A164324-10G A164324-1G A164324-25G A164324-5G
77110-792EA 208.06 USD InStock
77110-792 77110-794 77110-796 77110-798 77110-800
4'-Iodoacetanilide 95%
4'-Iodoacetanilide
Precaución: for Research Use Only


Formula: C₈H₈INO
MDL Number: MFCD00016352
CAS Number: 622-50-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty