Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-nitrobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A164438-100G A164438-25G A164438-5G
77110-802EA 48.4 USD InStock
77110-802 77110-804 77110-806
4-Chloro-2-nitrobenzonitrile 98%
4-Chloro-2-nitrobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClN₂O₂
MDL Number: MFCD00027398
CAS Number: 34662-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty