Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A164790-10G A164790-25G A164790-5G
77110-808EA 11.34 USD InStock
77110-808 77110-810 77110-812
3,4-Dichlorobenzonitrile 97%
3,4-Dichlorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃Cl₂N
MW: 172.01
MDL Number: MFCD00016379
CAS Number: 6574-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty