Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3,5-dimethylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A167339-1G A167339-250MG A167339-5G
77110-888EA 6.18 USD InStock
77110-888 77110-890 77110-892
4-Amino-3,5-dimethylbenzonitrile 97%
4-Amino-3,5-dimethylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₁₀N₂
MDL Number: MFCD07369841
CAS Number: 74896-24-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty