Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP-673451 98%

Supplier: AMBEED, INC
Warning

A171069-10MG A171069-1MG A171069-25MG A171069-5MG A171069-50MG
77111-010EA 56.65 USD InStock
77111-010 77111-012 77111-014 77111-016 77685-380
CP-673451 98%
CP-673451
Precaución: for Research Use Only


Formula: C₂₄H₂₇N₅O₂
MDL Number: MFCD11100329
CAS Number: 343787-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty