Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester 97%

Supplier: AMBEED, INC
Warning

A171976-1G A171976-250MG A171976-25G A171976-5G
77111-044EA 13.39 USD InStock
77111-044 77111-046 77111-048 77111-050
3-Methylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester 97%
3-Methylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₄
MDL Number: MFCD08461329
CAS Number: 534602-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty