Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)cinnamaldehyde 98%

Supplier: AMBEED, INC
Warning

A176684-100G A176684-1G A176684-250MG A176684-25G A176684-5G
77111-224EA 235.87 USD InStock
77111-224 77111-226 77111-228 77111-230 77111-232
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO
MDL Number: MFCD00007002
CAS Number: 6203-18-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty