Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NSC 66811 95%

Supplier: AMBEED, INC
Warning

A183021-10MG A183021-1MG A183021-25MG A183021-50MG A183021-5MG
77111-464EA 81.37 USD InStock
77111-464 77111-466 77111-468 77111-470 77111-472
NSC 66811 95%
NSC 66811
Precaución: for Research Use Only


Formula: C₂₃H₂₀N₂O
MW: 340.43 g/mol
MDL Number: MFCD08823776
CAS Number: 6964-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty