Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-fluoroisonicotinic acid 97%

Supplier: AMBEED, INC
Warning

A183174-100G A183174-10G A183174-1G A183174-25G A183174-5G
77111-486EA 184.37 USD InStock
77111-486 77111-488 77111-490 77111-492 77111-494
2-Chloro-3-fluoroisonicotinic acid 97%
2-Chloro-3-fluoroisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
MDL Number: MFCD04038325
CAS Number: 628691-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty