Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluorobenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A185017-100G A185017-10G A185017-250MG A185017-25G A185017-5G
77111-548EA 236.9 USD InStock
77111-548 77111-550 77111-552 77111-554 77111-556
3-Chloro-5-fluorobenzaldehyde 97%
3-Chloro-5-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄ClFO
MW: 158.56
MDL Number: MFCD02261734
CAS Number: 90390-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty