Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-cyano-4-fluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A186556-250MG A186556-5G A186556-10G
77111-598EA 7.21 USD InStock
77111-598 77111-600 77685-438
Methyl-3-cyano-4-fluorobenzoate 98%
Methyl-3-cyano-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₆FNO₂
MDL Number: MFCD06203915
CAS Number: 676602-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty