Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorophenyl isothiocyanate 96%

Supplier: AMBEED, INC
Danger

A193507-1G A193507-25G A193507-5G
77111-876EA 17.51 USD InStock
77111-876 77111-878 77111-880
2,6-Difluorophenyl isothiocyanate 96%
2,6-Difluorophenyl isothiocyanate
Precaución: for Research Use Only


Formula: C₇H₃F₂NS
MDL Number: MFCD00041047
CAS Number: 207974-17-2
UN: 1760
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty