Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethoxypyrimidine 97%

Supplier: AMBEED, INC
Warning

A194499-10G A194499-1G A194499-250MG A194499-25G A194499-5G
77111-926EA 19.57 USD InStock
77111-926 77111-928 77111-930 77111-932 77111-934
4,6-Dimethoxypyrimidine 97%
4,6-Dimethoxypyrimidine
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
MDL Number: MFCD00955844
CAS Number: 5270-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty