Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethoxy)anisole 98%

Supplier: AMBEED, INC
Warning

A194543-100G A194543-1G A194543-25G A194543-5G
77111-936EA 237.94 USD InStock
77111-936 77111-938 77111-940 77111-942
3-(Trifluoromethoxy)anisole 98%
3-(Trifluoromethoxy)anisole
Precaución: for Research Use Only


Formula: C₈H₇F₃O₂
MW: 192.14
MDL Number: MFCD01631504
CAS Number: 142738-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty