Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A195615-1G A195615-250MG A195615-25G A195615-5G
77111-968EA 25.75 USD InStock
77111-968 77111-970 77111-972 77111-974
3-Amino-4-methylphenylboronic acid 98%
3-Amino-4-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₁₀BNO₂
MW: 150.97 g/mol
MDL Number: MFCD01074640
CAS Number: 22237-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty