Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxy-N-(2,4-difluorophenyl)benzamide 98+%

Supplier: AMBEED, INC
Danger

A198314-10MG A198314-25MG A198314-50MG
77112-066EA 26.78 USD InStock
77112-066 77112-068 77112-070
4-Butoxy-N-(2,4-difluorophenyl)benzamide 98+%
4-Butoxy-N-(2,4-difluorophenyl)benzamide
Precaución: for Research Use Only


Formula: C₁₇H₁₇F₂NO₂
MDL Number: MFCD03243692
CAS Number: 433967-28-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty