Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-dichlorobenzotrifluoride 97%

Supplier: AMBEED, INC
Warning

A199806-100G A199806-10G A199806-1G A199806-250MG A199806-25G A199806-5G
77112-116EA 151.41 USD InStock
77112-116 77112-118 77112-120 77112-122 77112-124 77112-126
4-Bromo-3,5-dichlorobenzotrifluoride 97%
4-Bromo-3,5-dichlorobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₂BrCl₂F₃
MDL Number: MFCD00221020
CAS Number: 118754-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty