Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylpiperidine-4-carboxylic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A199875-100G A199875-10G A199875-1G A199875-5G
77112-130EA 69.01 USD InStock
77112-130 77112-132 77112-134 77112-136
1-Methylpiperidine-4-carboxylic acid hydrochloride 97%
1-Methylpiperidine-4-carboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₂
MDL Number: MFCD00190235
CAS Number: 71985-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty