Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(methylamino)benzamide 97%

Supplier: AMBEED, INC
Warning

A200590-100MG A200590-1G A200590-250MG A200590-5G
77112-160EA 41.21 USD InStock
77112-160 77112-162 77112-164 77112-166
5-Bromo-2-(methylamino)benzamide 97%
5-Bromo-2-(methylamino)benzamide
Precaución: for Research Use Only


Formula: C₈H₉BrN₂O
MW: 229.08 g/mol
MDL Number: MFCD19442206
CAS Number: 22721-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty