Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-o-xylene 97%

Supplier: AMBEED, INC
Warning

A204664-100G A204664-10G A204664-1G A204664-25G A204664-5G
77112-280EA 58.71 USD InStock
77112-280 77112-282 77112-284 77112-286 77112-288
4-Iodo-o-xylene 97%
4-Iodo-o-xylene


Formula: C₈H₉I
MDL Number: MFCD00040989
CAS Number: 31599-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty