Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,2'R)-4,4'-Disulfanediylbis(2-aminobutanoic acid) 96%

Supplier: AMBEED, INC
Warning

A206045-100MG A206045-1G A206045-250MG
77112-318EA 13.39 USD InStock
77112-318 77112-320 77112-322
(2R,2'R)-4,4'-Disulfanediylbis(2-aminobutanoic acid) 96%
(2R,2'R)-4,4'-Disulfanediylbis(2-aminobutanoic acid)
Precaución: for Research Use Only


Formula: C₈H₁₆N₂O₄S₂
MW: 268.35 g/mol
MDL Number: MFCD00068281
CAS Number: 6027-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty