Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-amino-4H-[1,2,4]triazole-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A209506-100G A209506-10G A209506-1G A209506-250MG A209506-5G
77112-404EA 215.26 USD InStock
77112-404 77112-406 77112-408 77112-410 77112-412
Ethyl-5-amino-4H-[1,2,4]triazole-3-carboxylate 98%
Ethyl-5-amino-4H-[1,2,4]triazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₅H₈N₄O₂
MDL Number: MFCD00186115
CAS Number: 63666-11-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty