Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-D-tyrosine 95%

Supplier: AMBEED, INC
Warning

A210830-100G A210830-10G A210830-1G A210830-25G A210830-5G
77112-434EA 219.39 USD InStock
77112-434 77112-436 77112-438 77112-440 77112-442
N-Carbobenzoxy-D-tyrosine 95%
N-Carbobenzoxy-D-tyrosine
Precaución: for Research Use Only


Formula: C₁₇H₁₇NO₅
MW: 315.33 g/mol
MDL Number: MFCD00063031
CAS Number: 64205-12-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty