Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-bromo-3-fluoro-5-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A216283-100G A216283-100MG A216283-10G A216283-1G A216283-250MG A216283-25G A216283-5G
77112-572EA 336.81 USD InStock
77112-572 77112-574 77112-576 77112-578 77112-580 77112-582 77112-584
2-bromo-3-fluoro-5-methylpyridine 98%
2-bromo-3-fluoro-5-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrFN
MDL Number: MFCD09909840
CAS Number: 34552-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty