Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-2-methylpropanenitrile 98%

Supplier: AMBEED, INC
Warning

A217776-1G A217776-250MG A217776-5G
77112-650EA 26.78 USD InStock
77112-650 77112-652 77112-654
2-(4-Bromophenyl)-2-methylpropanenitrile 98%
2-(4-Bromophenyl)-2-methylpropanenitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrN
MDL Number: MFCD02684219
CAS Number: 101184-73-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty