Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxymethylenemalononitrile 95%

Supplier: AMBEED, INC
Danger

A220054-100G A220054-25G A220054-500G A220054-5G
77112-716EA 55.63 USD InStock
77112-716 77112-718 77112-720 77112-722
Ethoxymethylenemalononitrile 95%
Ethoxymethylenemalononitrile
Precaución: for Research Use Only


Formula: C₆H₆N₂O
MDL Number: MFCD00001854
CAS Number: 123-06-8
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty