Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(((Benzyloxy)carbonyl)amino)malonic acid 97%

Supplier: AMBEED, INC
Warning

A221407-100MG A221407-1G A221407-250MG
77112-766EA 23.69 USD InStock
77112-766 77112-768 77112-770
2-(((Benzyloxy)carbonyl)amino)malonic acid 97%
2-(((Benzyloxy)carbonyl)amino)malonic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₆
MW: 253.21 g/mol
MDL Number: MFCD09841917
CAS Number: 76387-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty