Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluorophenylhydrazinium chloride 97%

Supplier: AMBEED, INC
Warning

A224677-100G A224677-10G A224677-25G A224677-5G
77112-820EA 99.91 USD InStock
77112-820 77112-822 77112-824 77112-826
3-Fluorophenylhydrazinium chloride 97%
3-Fluorophenylhydrazinium chloride
Precaución: for Research Use Only


Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD00012934
CAS Number: 2924-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty