Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tafamidis 98+%

Supplier: AMBEED, INC
Warning

A225692-5MG A225692-250MG A225692-1G
77112-830EA 31.83 USD InStock
77112-830 77112-832 77612-202 77685-618
Tafamidis 98+%
Tafamidis
Precaución: for Research Use Only


Formula: C₁₄H₇Cl₂NO₃
MW: 308.1 g/mol
MDL Number: MFCD16621109
CAS Number: 594839-88-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty