Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-fluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A230961-100G A230961-1G A230961-25G A230961-5G
77112-998EA 144.2 USD InStock
77112-998 77113-000 77113-002 77113-004
2-Chloro-3-fluorobenzoic acid 97%
2-Chloro-3-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
MDL Number: MFCD00973903
CAS Number: 102940-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty