Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mesityl-3a,4,5,6,7,7a-hexahydro-4,7-methanobenzo[d]isoxazole 98%

Supplier: AMBEED, INC
Warning

A239590-10MG A239590-1MG
77113-228EA 88.58 USD InStock
77113-228 77113-230
3-Mesityl-3a,4,5,6,7,7a-hexahydro-4,7-methanobenzo[d]isoxazole 98%
3-Mesityl-3a,4,5,6,7,7a-hexahydro-4,7-methanobenzo[d]isoxazole
Precaución: for Research Use Only


Formula: C₁₇H₂₁NO
MDL Number: MFCD09266779
CAS Number: 823218-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty