Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Difluoromethoxy)-3-fluorobenzonitrile 95%

Supplier: AMBEED, INC
Warning

A239793-100MG A239793-1G A239793-250MG A239793-5G
77113-232EA 14.42 USD InStock
77113-232 77113-234 77113-236 77113-238
4-(Difluoromethoxy)-3-fluorobenzonitrile 95%
4-(Difluoromethoxy)-3-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₈H₄F₃NO
MDL Number: MFCD18394957
CAS Number: 954388-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty