Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A242501-25G A242501-5G
77113-276EA 56.65 USD InStock
77113-276 77113-278
3-Iodobenzyl alcohol 98%
3-Iodobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇IO
MW: 234.04
Boiling Pt: 252
Density: 1.84
MDL Number: MFCD00004635
CAS Number: 57455-06-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty