Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycyl-L-glutamine 97%

Supplier: AMBEED, INC
Warning

A243057-10G A243057-25G A243057-5G
77113-280EA 40.18 USD InStock
77113-280 77113-282 77113-284
Glycyl-L-glutamine 97%
Glycyl-L-glutamine
Precaución: for Research Use Only


Formula: C₇H₁₃N₃O₄
MW: 203.198 g/mol
MDL Number: MFCD00077124
CAS Number: 13115-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty