Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IWP-2 ≥98%

Supplier: AMBEED, INC
Warning

A250953-10MG A250953-50MG A250953-5MG
77113-452EA 67.99 USD InStock
77113-452 77113-454 77113-456 77113-458
IWP-2 ≥98%
IWP-2
Precaución: for Research Use Only


Formula: C₂₂H₁₈N₄O₂S₃
MDL Number: MFCD12912418
CAS Number: 686770-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty