Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methyl-5-pyridineboronic acid 97%

Supplier: AMBEED, INC
Warning

A255322-1G A255322-250MG A255322-5G
77113-528EA 15.45 USD InStock
77113-528 77113-530 77113-532
2-Fluoro-6-methyl-5-pyridineboronic acid 97%
2-Fluoro-6-methyl-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BFNO₂
MW: 154.9400 g/mol
MDL Number: MFCD03411561
CAS Number: 904326-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty