Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 2-carbamoylpiperidine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A256180-100MG A256180-250MG
77113-564EA 50.47 USD InStock
77113-564 77113-566
(R)-tert-Butyl 2-carbamoylpiperidine-1-carboxylate 98%
(R)-tert-Butyl 2-carbamoylpiperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD03840195
CAS Number: 848488-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty